Skip to Content
Merck
All Photos(1)

Key Documents

422959

Sigma-Aldrich

3,5,5-Trimethylhexanoyl chloride

98%

Synonym(s):

Isononanoyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CCH2CH(CH3)CH2COCl
CAS Number:
Molecular Weight:
176.68
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.436 (lit.)

bp

188-190 °C (lit.)

density

0.93 g/mL at 25 °C (lit.)

SMILES string

CC(CC(Cl)=O)CC(C)(C)C

InChI

1S/C9H17ClO/c1-7(5-8(10)11)6-9(2,3)4/h7H,5-6H2,1-4H3

InChI key

GEKPNPPFAYJZRD-UHFFFAOYSA-N

General description

3,5,5-Trimethylhexanoyl chloride (isononanoyl chloride) is an acid chloride. Its has been reported to be synthesized by the chlorination of isononanoic acid with thionyl chloride.[1]

Application

3,5,5-Trimethylhexanoyl chloride (isononanoyl chloride) may be used:
  • As a reagent in the synthesis of barbituric acid analogs[2]
  • As a starting material in the synthesis of α-N-fattyacyl colistin nonapeptide derivative.[1]
  • As a reactant in the synthesis of 3,5,5-trimethylhexanamide.[3]

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Met. Corr. 1 - Skin Corr. 1A - Skin Sens. 1

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

284.0 °F - closed cup

Flash Point(C)

140 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemical synthesis, isolation and characterization of a-N-fattyacyl colistin nonapeptide with special reference to the correlation between antimicrobial activity and carbon number of fattyacyl moiety.
Chihara S, et al.
Agricultural and Biological Chemistry, 38(3), 521-529 (1974)
Outer-Sphere Coordination Chemistry: Amido-Ammonium Ligands as Highly Selective Tetrachloridozinc (II) ate Extractants.
Turkington JR, et al.
Inorganic Chemistry, 51(23), 12805-12819 (2012)
Yong-Chul Jeong et al.
Molecules (Basel, Switzerland), 20(3), 3582-3627 (2015-02-25)
The synthesis, tautomerism and antibacterial activity of novel barbiturates is reported. In particular, 3-acyl and 3-carboxamidobarbiturates exhibited antibacterial activity, against susceptible and some resistant Gram-positive strains of particular interest is that these systems possess amenable molecular weight, rotatable bonds and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service