Skip to Content
Merck
All Photos(1)

Key Documents

422363

Sigma-Aldrich

Diketene

contains copper sulfate as stabilizer

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4O2
CAS Number:
Molecular Weight:
84.07
Beilstein:
104541
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

2.9 (vs air)

vapor pressure

7.9 mmHg ( 20 °C)

form

liquid

autoignition temp.

590 °F

contains

copper sulfate as stabilizer

refractive index

n20/D 1.439 (lit.)

bp

69-70 °C/100 mmHg (lit.)

density

1.09 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C=C1CC(=O)O1

InChI

1S/C4H4O2/c1-3-2-4(5)6-3/h1-2H2

InChI key

WASQWSOJHCZDFK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Employed in the convenient preparation of acetoacetic acid derivatives and heterocycles.

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

93.2 °F - closed cup

Flash Point(C)

34 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ahmad Shaabani et al.
Journal of combinatorial chemistry, 11(3), 375-377 (2009-03-14)
In this work, 1,2,3,4,5,8-hexahydro-1,3,7-trimethyl-2,4-dioxopyrido[2,3-d]pyrimidine-6-carboxamide derivatives were synthesized in a simple and efficient method from the four-component condensation reaction of diketene, an aliphatic or aromatic amine, an aromatic aldehyde, and 6-amino-1,3-dimethyluracil in the presence of a catalytic amount of p-toluenesulfonic acid
Nazariy T Pokhodylo et al.
Journal of combinatorial chemistry, 11(3), 481-485 (2009-04-23)
A convenient synthetic protocol was elaborated for creation of combinatorial libraries of 1-(R(1)-phenyl)-5-methyl-N-R(2)-1H-1,2,3-triazole-4-carboxamides. As starting materials, commercially available or readily prepared azides, amines, and diketene were selected for the reaction which has proceeded in a one-pot system with high yields
S Breda et al.
The journal of physical chemistry. A, 116(9), 2131-2140 (2012-01-26)
Diketene (C(4)H(4)O(2)) monomers were isolated in cryogenic Ar (15 K) and Xe (30 K) matrices. The infrared (IR) spectra of the freshly deposited matrices show that diketene monomers exclusively adopt the 4-methylene-oxetan-2-one form. In situ photochemical transformations of diketene were
Identification of essential amino acid residues in clostridium histolyticum collagenase using chemical modification reactions.
M D Bond et al.
Biochemical and biophysical research communications, 102(1), 243-249 (1981-09-16)
A Jonas et al.
Biochemistry, 24(14), 3508-3513 (1985-07-02)
Discoidal complexes of human apolipoprotein A-I-egg phosphatidylcholine-cholesterol were prepared by the sodium cholate dialysis procedure and were reacted to varying extents with the amino group reagents citraconic anhydride, diketene, and formaldehyde in the presence of sodium borohydride. Modification of positive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service