Skip to Content
Merck
All Photos(1)

Key Documents

42000301

Sigma-Aldrich

PRONOVA® UP MVM

medium viscosity sodium alginate, viscosity >200 mPa.s, 1 %(20 °C)(low), mannuronate monomer units ≥50 %

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$791.00
10 G
$1,580.00

$791.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$791.00
10 G
$1,580.00

About This Item

UNSPSC Code:
12162002
NACRES:
NA.25

$791.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

form

powder

mol wt

Mw >200 kDa (approximate)

composition

mannuronate monomer units, ≥50%
G/M ratio, ≤1

impurities

≤100 CFU/g Total viable count
≤100 EU/g Endotoxins

pH

5.5-8.5 (20 °C, 1% in water)

viscosity

>200 mPa.s, 1 %(20 °C) (
low)

General description

PRONOVA UP MVM is a medium viscosity (>200 mPas) sodium alginate where minimum 50% of the monomer units are mannuronate.

Application

Examples of Pronova Alginates applications include: encapsulation of living cells and therapeutic proteins for cell therapy medications, cryoprotectant, bone putty binder for tissue engineering, 3D bioprinting, foams and cell tissue scaffolds, hydrogels, advanced drug delivery and implants, wound management, and medical devices.

Legal Information

Product of Dupont Nutrition USA, Inc.
PRONOVA is a registered trademark of International N&H USA, Inc.

Disclaimer

NOT FOR USE IN HUMANS

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Engineered ECMs enhance immune therapy in cancer treatment by supporting cells and tissues and modulating immune response. They improve immune cell maturation, expansion, and regulation through biomaterial manipulation, acting as frameworks or carriers for enhanced tumor immunotherapy.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service