Skip to Content
Merck
All Photos(1)

Key Documents

419605

Sigma-Aldrich

1-Methyl-1-propenylmagnesium bromide solution

0.5 M in THF

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$127.00

$127.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
100 ML
$127.00

About This Item

Linear Formula:
CH3CH=C(CH3)MgBr
CAS Number:
Molecular Weight:
159.31
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$127.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

reaction suitability

reaction type: Grignard Reaction

concentration

0.5 M in THF

bp

67 °C

density

0.934 g/mL at 25 °C

SMILES string

C\C=C(/C)[Mg]Br

InChI

1S/C4H7.BrH.Mg/c1-3-4-2;;/h3H,1-2H3;1H;/q;;+1/p-1

InChI key

FEJKWDUHNFCDRZ-UHFFFAOYSA-M

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3 - Water-react 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

-14.8 °F - closed cup

Flash Point(C)

-26 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service