Skip to Content
Merck
All Photos(3)

Key Documents

419303

Sigma-Aldrich

Sodium carboxymethyl cellulose

average Mw ~250,000, degree of substitution 0.9

Synonym(s):

Carboxymethylcellulose sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$93.70
1 KG
$200.00

$93.70

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$93.70
1 KG
$200.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

$93.70

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

form

powder

autoignition temp.

698 °F

mol wt

average Mw ~250,000

extent of labeling

0.9 carboxymethyl groups per anhydroglucose unit

mp

270 °C (dec.)

SMILES string

[Na].OC(C(O)C(O)C=O)C(O)CO.OC(=O)C

InChI

1S/C6H12O6.C2H4O2.Na/c7-1-3(9)5(11)6(12)4(10)2-8;1-2(3)4;/h1,3-6,8-12H,2H2;1H3,(H,3,4);

InChI key

DPXJVFZANSGRMM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sodium carboxymethyl cellulose (CMC) is a class of linear structured cellulose which consists of polysaccharide composing the fibrous tissue of plants.[1][2]

Application

CMC can be used for a wide range of application such as:
  • food processing, textiles
  • detergents, and drugs[1]
  • a stabilizer in the preparation of nanoparticles[3]
  • a binder for the fabrication of electrodes[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alan Ransil et al.
Nature communications, 12(1), 6494-6494 (2021-11-13)
Sodium trisilicate waterglass is an earth-abundant inorganic adhesive which binds to diverse materials and exhibits extreme chemical and temperature stability. Here we demonstrate the use of this material as an electrode binder in a lay-up based manufacturing system to produce
Sodium carboxymethyl cellulose a potential binder for Si negative electrodes for Li-ion batteries
Li J, et al.
Electrochemical and Solid-State Letters, 10(2), A17-A20 (2007)
Lisa Miclotte et al.
Frontiers in microbiology, 11, 577474-577474 (2020-12-01)
The use of additives in food products has become an important public health concern. In recent reports, dietary emulsifiers have been shown to affect the gut microbiota, contributing to a pro-inflammatory phenotype and metabolic syndrome. So far, it is not
In situ synthesis of polyaniline/sodium carboxymethyl cellulose nanorods for high-performance redox supercapacitors
Peng H, et al.
Journal of Power Sources, 211, 40-45 (2012)
Silver nanoparticles as pigments for water-based ink-jet inks
Magdassi S, et al.
Chemistry of Materials, 15(11), 2208-2217 (2003)

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service