Skip to Content
Merck
All Photos(1)

Documents

417645

Sigma-Aldrich

2-Chloroethyl phenyl sulfone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5SO2CH2CH2Cl
CAS Number:
Molecular Weight:
204.67
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

bp

168-174 °C/7 mmHg (lit.)

mp

55-57 °C (lit.)

SMILES string

ClCCS(=O)(=O)c1ccccc1

Gene Information

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service