Skip to Content
Merck
All Photos(1)

Key Documents

417483

Sigma-Aldrich

6-Bromochromone-2-carboxylic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H5BrO4
CAS Number:
Molecular Weight:
269.05
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

mp

~265 °C (dec.) (lit.)

SMILES string

OC(=O)C1=CC(=O)c2cc(Br)ccc2O1

InChI

1S/C10H5BrO4/c11-5-1-2-8-6(3-5)7(12)4-9(15-8)10(13)14/h1-4H,(H,13,14)

InChI key

QSBZDBNPXSVVHH-UHFFFAOYSA-N

General description

6-Bromochromone-2-carboxylic acid is a chromone derivative. Chromones are cyclic compounds containing nine carbon atoms with sp2 hybridization. They are commonly referred as 1,4-benzapyranes or γ-benzopyrones.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The standard molar enthalpies of formation of chromone-3-carboxylic acid, 6-methylchromone-2-carboxylic acid, and 6-methyl-4-chromanone determined by micro-combustion calorimetry.
Flores H, et al.
Journal of Thermal Analysis and Calorimetry, 117(1), 433-437 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service