Skip to Content
Merck
All Photos(1)

Key Documents

416274

Sigma-Aldrich

N-Phenylphthalimide

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H9NO2
CAS Number:
Molecular Weight:
223.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

form

powder

mp

204-207 °C (lit.)

SMILES string

O=C1N(c2ccccc2)C(=O)c3ccccc13

InChI

1S/C14H9NO2/c16-13-11-8-4-5-9-12(11)14(17)15(13)10-6-2-1-3-7-10/h1-9H

InChI key

MFUPLJQNEXUUDW-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hiroko Sano et al.
Chemical & pharmaceutical bulletin, 52(8), 1021-1022 (2004-08-12)
Several N-substituted phenylphthalimide and phenylhomophthalimide derivatives with cyclooxygenase (COX)-inhibitory activity were prepared during structural development studies based on thalidomide as a lead compound. Substituent effects on the subtype selectivity were investigated.
R Shimazawa et al.
Bioorganic & medicinal chemistry letters, 9(4), 559-562 (1999-03-31)
A novel series of nonpeptide small-molecular dipeptidyl peptidase IV (DPP-IV) inhibitors with an N-phenylphthalimide skeleton has been developed. Some of the compounds, including 4-amino-(2,6-dimethylphenyl)phthalimides (7), 4- and 5-hydroxy-(2,6-diethylphenyl)phthalimide (11 and 14), 4-hydroxy-(2,6-diisopropylphenyl)phthalimide (12), and thiocarbonyl analogs of (2,6-diisopropylphenyl)phthalimide and their
H Takahashi et al.
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan, 120(10), 909-921 (2000-11-18)
The studies on both structure-activity relationship study and identification of the target enzyme of novel nonpeptide aminopeptidase inhibitors with cyclic imide skeleton are reviewed. Some N-phenylphthalimide or N-phenylhomophthalimide derivative showed potent protease inhibitory activity in an assay system using human
The oxidative degradation of imide polymers. I: Ozonolysis of a model compound, N-phenylphthalimide.
Barman S, et al.
Polym. Eng. Sci., 34(4), 279-284 (1994)
Y Shibata et al.
Chemical & pharmaceutical bulletin, 44(1), 156-162 (1996-01-01)
Phenylphthalimides (2-phenyl-1H-isoindole-1,3-diones) were prepared and their effects on tumor necrosis factor alpha (TNF-alpha) production by human leukemia cell line HL-60 stimulated with 12-O-tetradecanoylphorbol-13-acetate (TPA) were examined. An analysis of the structure-activity relationships of the phenylphthalimides indicated that potent enhancing activity

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service