Skip to Content
Merck
All Photos(1)

Key Documents

415553

Sigma-Aldrich

D-Fructose-1-13C

99 atom % 13C

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$744.00
500 MG
$1,200.00

$744.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Select a Size

Change View
250 MG
$744.00
500 MG
$1,200.00

About This Item

Empirical Formula (Hill Notation):
13CC5H12O6
CAS Number:
Molecular Weight:
181.15
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.12

$744.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

isotopic purity

99 atom % 13C

Assay

≥99% (HPLC)

form

powder

optical activity

[α]20/D -93.5°, c = 2 in H2O (trace NH4OH)

mp

119-122 °C (dec.) (lit.)

mass shift

M+1

SMILES string

OC[C@H]1OC(O)([13CH2]O)[C@@H](O)[C@@H]1O

InChI

1S/C6H12O6/c7-1-3-4(9)5(10)6(11,2-8)12-3/h3-5,7-11H,1-2H2/t3-,4-,5+,6?/m1/s1/i2+1

InChI key

RFSUNEUAIZKAJO-BORCATHGSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raphael Tiziani et al.
Scientific reports, 10(1), 15970-15970 (2020-10-01)
Plant roots are able to exude vast amounts of metabolites into the rhizosphere in response to phosphorus (P) deficiency. Causing noteworthy costs in terms of energy and carbon (C) for the plants. Therefore, it is suggested that exudates reacquisition by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service