Skip to Content
Merck
All Photos(1)

Documents

413615

Sigma-Aldrich

Barium iodide

AnhydroBeads, −10 mesh, 99.995% trace metals basis

Synonym(s):

Barium diiodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BaI2
CAS Number:
Molecular Weight:
391.14
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

product line

AnhydroBeads

Assay

99.995% trace metals basis

form

beads

impurities

≤55.0 ppm Trace Metal Analysis

particle size

−10 mesh

mp

740 °C (lit.)

density

5.15 g/mL at 25 °C (lit.)

SMILES string

I[Ba]I

InChI

1S/Ba.2HI/h;2*1H/q+2;;/p-2

InChI key

SGUXGJPBTNFBAD-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Reagent for facile synthesis of poly(phenylcarbyne), a precursor to diamond-like carbon.

Legal Information

AnhydroBeads is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gurdip K Daffu et al.
Protein engineering, design & selection : PEDS, 28(11), 501-506 (2015-08-06)
The catalytic bioscavenger phosphotriesterase (PTE) is experimentally an effective antidote for organophosphate poisoning. We are interested in the molecular engineering of this enzyme to confer additional functionality, such as improved in vivo longevity. To this aim, we developed PTE cysteine
Rieke, R.D.; Chen. T-A.
Chemistry of Materials, 6, 576-576 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service