Skip to Content
Merck
All Photos(1)

Key Documents

413194

Sigma-Aldrich

N-(1-Acetyl-2,2,6,6-tetramethyl-4-piperidinyl)-2-dodecylsuccinimide

Synonym(s):

1-(1-Acetyl-2,2,6,6-tetramethyl-4-piperidinyl)-3-dodecyl-2,5-pyrrolidinedione, 1-Acetyl-4-(3-dodecyl-2,5-dioxo-1-pyrrolidinyl)-2,2,6,6-tetramethylpiperidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H48N2O3
CAS Number:
Molecular Weight:
448.68
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

refractive index

n20/D 1.495 (lit.)

bp

222 °C (lit.)

density

0.977 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCCC1CC(=O)N(C2CC(C)(C)N(C(C)=O)C(C)(C)C2)C1=O

InChI

1S/C27H48N2O3/c1-7-8-9-10-11-12-13-14-15-16-17-22-18-24(31)28(25(22)32)23-19-26(3,4)29(21(2)30)27(5,6)20-23/h22-23H,7-20H2,1-6H3

InChI key

VXTRPEFUPWORNH-UHFFFAOYSA-N

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tarick J El-Baba et al.
Rapid communications in mass spectrometry : RCM, 28(11), 1175-1184 (2014-04-25)
New inlet and vacuum ionization methods provide advantages of specificity, simplicity and speed for the analysis of synthetic polymers and polymer additives directly from surfaces such as fibers using mass spectrometry (MS) on different commercial mass spectrometers (Waters SYNAPT G2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service