Skip to Content
Merck
All Photos(1)

Key Documents

412848

Sigma-Aldrich

Dodecyl and tetradecyl glycidyl ethers

technical grade

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical grade

form

liquid

eq. wt.

≥0.32

refractive index

n20/D 1.447 (lit.)

density

0.89 g/mL at 25 °C (lit.)

functional group

ether

SMILES string

CCCCCCCCCCCCOCC1CO1.CCCCCCCCCCCCCOCC2CO2.CCCCCCCCCCCCCCOCC3CO3

InChI

1S/C17H34O2.C16H32O2.C15H30O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-18-15-17-16-19-17;1-2-3-4-5-6-7-8-9-10-11-12-13-17-14-16-15-18-16;1-2-3-4-5-6-7-8-9-10-11-12-16-13-15-14-17-15/h17H,2-16H2,1H3;16H,2-15H2,1H3;15H,2-14H2,1H3

InChI key

CKNWGKWITWNVEY-UHFFFAOYSA-N

General description

Dodecyl and tetradecyl glycidyl ethers participates in the synthesis of high molecular weight poly(ethylene oxide)-b-poly(alkylglycidyl ether) diblock copolymers.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Core-corona nanoparticles formed by high molecular weight poly (ethylene oxide)-b-poly (alkylglycidyl ether) diblock copolymers.
Petrov P, et al.
Polymer, 43(25), 6641-6651 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service