Skip to Content
Merck
All Photos(1)

Key Documents

412791

Sigma-Aldrich

N,O-Di-Boc-hydroxylamine

97%

Synonym(s):

N,O-Bis(tert-butoxycarbonyl)hydroxylamine, tert-Butyl N-(tert-butoxycarbonyloxy)carbamate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$129.00

$129.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$129.00

About This Item

Linear Formula:
(CH3)3COCONHOCOOC(CH3)3
CAS Number:
Molecular Weight:
233.26
Beilstein:
1794022
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$129.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

mp

67-70 °C (lit.)

functional group

amine

storage temp.

−20°C

SMILES string

CC(C)(C)OC(=O)NOC(=O)OC(C)(C)C

InChI

1S/C10H19NO5/c1-9(2,3)14-7(12)11-16-8(13)15-10(4,5)6/h1-6H3,(H,11,12)

InChI key

AGOSGCWATIJZHQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N,O-Di-Boc-hydroxylamine (N,O-Bis(tert-butoxycarbonyl)hydroxylamine) may be used for the synthesis of the following:
  • 5-lipoxygenase inhibitor LY280810[1]
  • hydroxylamines[1]
  • hydroxamic acids[1],[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The use of N, O-bis (tert-butoxycarbonyl)-hydroxylamine in the synthesis of N-hydroxylamines and hydroxamic acids.
Staszak MA and Doecke CW.
Tetrahedron Letters, 35(23), 6021-6024 (1994)
A facile synthesis of N, O-bis (tert-butoxycarbonyl)-hydroxylamine.
Staszak MA and Doecke CW.
Tetrahedron Letters, 34(44), 7043-7044 (1993)
A convenient method for the preparation of hydroxamic acids.
Reddy AS, et al.
Tetrahedron Letters, 41(33), 6285-6288 (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service