Skip to Content
Merck
All Photos(1)

Key Documents

412414

Sigma-Aldrich

PAMAM dendrimer, ethylenediamine core, generation 2.5 solution

10 wt. % in methanol

Synonym(s):

PAMAM-G 2.5

Sign Into View Organizational & Contract Pricing

Select a Size

2.5 G
$315.00

$315.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
2.5 G
$315.00

About This Item

Linear Formula:
[NH2(CH2)2NH2]:(G=2.5);dendri PAMAM(NHCH2CH2COONa)32
CAS Number:
Molecular Weight:
6265.60
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

$315.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

description

sodium carboxylate surface groups

form

solution

mol wt

generation 2.5

feature

core type ethylenediamine core (2-carbon core)

concentration

10 wt. % in methanol

no. Surface Groups

32

refractive index

n20/D 1.366

bp

65 °C

density

0.86 g/mL at 25 °C

storage temp.

2-8°C

Physical form

Contains 32 surface carboxylate groups

Legal Information

Manufactured by Dendritech®, Inc.
Dendritech is a registered trademark of Dendritech, Inc.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service