Skip to Content
Merck
All Photos(1)

Key Documents

412333

Sigma-Aldrich

Soybean oil, epoxidized acrylate

Synonym(s):

ESOA

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
$378.00

$378.00

PRICE WITHOUT NATIONAL TAXES

Available to ship onApril 07, 2025Details


Request a Bulk Order

Select a Size

Change View
1 L
$378.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

$378.00

PRICE WITHOUT NATIONAL TAXES

Available to ship onApril 07, 2025Details


Request a Bulk Order

form

viscous liquid

refractive index

n20/D 1.484 (lit.)

density

1.04 g/mL at 25 °C (lit.)

General description

Epoxidized soybean oil acrylate (ESOA) can be synthesized by reacting acrylic acid with epoxidised soybean oil. [1]

Application

  • Smart Biomedical Scaffolds: Application in 4D printing of smart biomedical scaffolds using novel soybean oil epoxidized acrylate, demonstrating the material′s adaptability and function in medical implant technologies (Miao, Zhu, Castro, Nowicki, Zhou, Cui, 2016).
  • UV-Curable Phosphorous-Containing Resins: Synthesis of UV-curable phosphorous-containing acrylated epoxidized soybean oil-based resins aimed at enhancing fire resistance and durability of the resulting polymers (Hu, Jia, Shang, Zhang, Feng, Liu, 2019).
  • Slow-Release Fertilizer: Development of slow-release fertilizers by coating urea with acrylate epoxidized soybean oil, aiming to improve nutrient use efficiency and reduce environmental impact (Jiang, Duan, Ma, Song, Xie, Liu, 2024).
  • High-Performance Epoxy Resins: Creation of high-performance, bio-based epoxy acrylate resins from soybean oil for use in UV-curable coatings, offering a sustainable alternative to conventional petrochemical-derived resins (Wu, Hu, Tang, Zhang, Wang, 2018).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, characterization, and kinetics study of thermal decomposition of epoxidized soybean oil acrylate.
Behera D and Banthia AK
Journal of Applied Polymer Science, 109(4, 2583-2590 (2008)
Dibakar Mondal et al.
Journal of the mechanical behavior of biomedical materials, 104, 103653-103653 (2020-03-17)
In this study, single filaments of acrylated epoxidized soybean oil (AESO)/polyethylene glycol diacrylate (PEGDA)/nanohydroxyapatite (nHA)-based nanocomposites intended for bone defect repair have displayed significant improvement of their mechanical properties when extruded through smaller needle gauges before UV curing. These nanocomposite

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. Hello, I'm wondering what the functionality of this product is, e.g. how many epoxides/acrylates can I expect per unit volume or mass? Is this something you report or something we need to determine empirically?

    1 answer
    1. Soybean oil, epoxidized acrylate, is fully converted to the acrylate form with 1-2 acrylates per molecule. A structure is not available for this product. There are three epoxy groups on the molecule, and they react with the acrylate to form an epoxidized soybean oil acrylate.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service