Skip to Content
Merck
All Photos(1)

Key Documents

411884

Sigma-Aldrich

Phenylacetylene-d

99 atom % D

Synonym(s):

(Ethynyl-d)benzene

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$1,110.00

$1,110.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Select a Size

Change View
5 G
$1,110.00

About This Item

Linear Formula:
C6H5C≡CD
CAS Number:
Molecular Weight:
103.14
Beilstein:
2038975
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

$1,110.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

isotopic purity

99 atom % D

Assay

99% (CP)

form

liquid

refractive index

n20/D 1.548 (lit.)

bp

142-144 °C (lit.)

density

0.939 g/mL at 25 °C

mass shift

M+1

SMILES string

[2H]C#Cc1ccccc1

InChI

1S/C8H6/c1-2-8-6-4-3-5-7-8/h1,3-7H/i1D

InChI key

UEXCJVNBTNXOEH-MICDWDOJSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

87.8 °F - closed cup

Flash Point(C)

31 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Organometallics, 11, 2181-2181 (1992)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service