Skip to Content
Merck
All Photos(2)

Key Documents

411345

Sigma-Aldrich

N-Trityl-L-serine methyl ester

99%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$534.00

$534.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$534.00

About This Item

Linear Formula:
HOCH2CH[NHC(C6H5)3]CO2CH3
CAS Number:
Molecular Weight:
361.43
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

$534.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

99%

optical activity

[α]20/D +31°, c = 1 in methanol

reaction suitability

reaction type: solution phase peptide synthesis

mp

148-150 °C (lit.)

application(s)

peptide synthesis

SMILES string

COC(=O)[C@H](CO)NC(c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C23H23NO3/c1-27-22(26)21(17-25)24-23(18-11-5-2-6-12-18,19-13-7-3-8-14-19)20-15-9-4-10-16-20/h2-16,21,24-25H,17H2,1H3/t21-/m0/s1

InChI key

LXAWQKKSNNYYEK-NRFANRHFSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service