Skip to Content
Merck
All Photos(2)

Key Documents

411167

Sigma-Aldrich

Bisphenol A glycerolate (1 glycerol/phenol) diacrylate

contains MEHQ as inhibitor

Synonym(s):

2,2-Bis[4-(3-acryloyloxy-2-hydroxypropoxy)phenyl]propane, 2,2-Bis[acryloyloxy(2′-hydroxypropyloxy)phenyl]propane, Bisphenol A glycerolate diacrylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H32O8
CAS Number:
Molecular Weight:
484.54
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

contains

MEHQ as inhibitor
250-500 ppm MEHQ as inhibitor

Quality Level

refractive index

n20/D 1.557 (lit.)

viscosity

2000-4000 cP(65 °C)(lit.)

acid number

<5 mg KOH/g

density

1.18 g/mL at 25 °C (lit.)

SMILES string

CC(C)(c1ccc(OCC(O)COC(=O)C=C)cc1)c2ccc(OCC(O)COC(=O)C=C)cc2

InChI

1S/C27H32O8/c1-5-25(30)34-17-21(28)15-32-23-11-7-19(8-12-23)27(3,4)20-9-13-24(14-10-20)33-16-22(29)18-35-26(31)6-2/h5-14,21-22,28-29H,1-2,15-18H2,3-4H3

InChI key

VZTQQYMRXDUHDO-UHFFFAOYSA-N

Application

UV curable cross-linking agent. Component of curable inks, coatings, varnishes and lacquers.

Features and Benefits

High viscosity, non-volatile, difunctional acrylate. Gives fast cure speeds, low odor and low volatility.

Physical form

epoxy acrylate oligomer

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D L Holness et al.
Contact dermatitis, 28(4), 216-219 (1993-04-01)
Epoxy resin of the bisphenol A type (ERBA) is a common cause of occupational contact dermatitis. 167 patients with a history of potential exposure were patch tested with epoxy resin (molecular weight 340) (ERBAM) 1% in pet. and with a
B Björkner
Acta dermato-venereologica, 61(1), 7-10 (1981-01-01)
One commonly used prepolymer in ultraviolet (UV) curing inks is epoxy acrylate. Of 6 men with dermatitis contracted from UV-curing inks, 2 had positive patch test reaction to epoxy acrylate. None reacted to the chemically related bisphenol A dimethacrylate. The
L Kanerva et al.
Contact dermatitis, 14(2), 80-84 (1986-02-01)
A dental assistant developed sensitivity to dental restorative materials within 3 months of starting to use them. They contained the epoxy acrylate BIS-GMA, which is the most commonly used dimethacrylate monomer in dental composite restorations. She was positive to a
Han N Lee et al.
American journal of contact dermatitis : official journal of the American Contact Dermatitis Society, 13(3), 108-115 (2002-08-08)
The study's objective was 2-fold: first, to evaluate the potential cross-reactivity between Bis-A epoxy resins and epoxy acrylates and second, to study the cross reactivity between Bis-A epoxy resins and newer Bis-F epoxy resins in patients with allergic contact dermatitis
Li Li et al.
Electrophoresis, 33(16), 2591-2597 (2012-08-18)
Manufacturing materials are an essential element for the fabrication of microfluidic chips. PDMS, the most widely used polymeric material, is associated with apparent disadvantages such as hydrophobic nature, while other materials also suffer from some limitations. In this paper, a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service