Skip to Content
Merck
All Photos(1)

Key Documents

411094

Sigma-Aldrich

1,1-Dichlorosilacyclobutane

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$211.00

$211.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 ML
$211.00

About This Item

Empirical Formula (Hill Notation):
C3H6Cl2Si
CAS Number:
Molecular Weight:
141.07
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$211.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.464 (lit.)

bp

113-115 °C (lit.)

density

1.19 g/mL at 25 °C (lit.)

SMILES string

Cl[Si]1(Cl)CCC1

InChI

1S/C3H6Cl2Si/c4-6(5)2-1-3-6/h1-3H2

InChI key

PASYEMKYRSIVTP-UHFFFAOYSA-N

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - Water-react 1

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

68.0 °F - closed cup

Flash Point(C)

20 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service