Skip to Content
Merck
All Photos(2)

Key Documents

408948

Sigma-Aldrich

Isodecyl methacrylate

95%

Synonym(s):

Bisomer IDMA

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
$151.00

$151.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 L
$151.00

About This Item

Linear Formula:
H2C=C(CH3)CO2C10H21
CAS Number:
Molecular Weight:
226.36
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$151.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

95%

form

liquid

contains

150 ppm monomethyl ether hydroquinone as inhibitor

refractive index

n20/D 1.443 (lit.)

bp

126 °C/10 mmHg (lit.)

density

0.878 g/mL at 25 °C (lit.)

SMILES string

CC(C)CCCCCCCOC(=O)C(C)=C

InChI

1S/C14H26O2/c1-12(2)10-8-6-5-7-9-11-16-14(15)13(3)4/h12H,3,5-11H2,1-2,4H3

InChI key

COCLLEMEIJQBAG-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service