Skip to Content
Merck
All Photos(1)

Key Documents

408905

Sigma-Aldrich

Hexyl acrylate

98%, contains 100 ppm hydroquinone as inhibitor

Synonym(s):

Acrylic Acid Hexyl Ester, Hexyl 2-propenoate, hexylacrylate, n-Hexyl acrylate

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
$52.10
100 ML
$146.00

$52.10


Estimated to ship onMarch 22, 2025


Request a Bulk Order

Select a Size

Change View
25 ML
$52.10
100 ML
$146.00

About This Item

Linear Formula:
H2C=CHCO2(CH2)5CH3
CAS Number:
Molecular Weight:
156.22
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$52.10


Estimated to ship onMarch 22, 2025


Request a Bulk Order

Quality Level

Assay

98%

form

liquid

contains

100 ppm hydroquinone as inhibitor

refractive index

n20/D 1.428 (lit.)

bp

88-90 °C/24 mmHg (lit.)

density

0.888 g/mL at 25 °C (lit.)

SMILES string

CCCCCCOC(=O)C=C

InChI

1S/C9H16O2/c1-3-5-6-7-8-11-9(10)4-2/h4H,2-3,5-8H2,1H3

InChI key

LNMQRPPRQDGUDR-UHFFFAOYSA-N

General description

Poly(hexyl acrylate-co-1,4-butanediol diacrylate-co-[2-(acryloyloxy)ethyl]trimethyl ammonium chloride) can be prepared by the photopolymeirzation of hexyl acrylate and several other components. These polyacrylate-based monolithics were designed to be used for capillary electrochromatography (CEC). [1] Mutagenicity of this acrylate ester was evaluated by the Salmonella-microsome test. [2]

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

154.4 °F - closed cup

Flash Point(C)

68 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Non-mutagenicity of 27 aliphatic acrylate esters in the Salmonella-microsome test
Waegemaekers THJM, et al.
Mutation Research. Genetic Toxicology and Environmental Mutagenesis, 137(2-3), 95-102 (1984)
Braden L Leigh et al.
ACS applied materials & interfaces, 9(37), 31488-31496 (2017-08-26)
Cochlear Implants (CIs) suffer from limited tonal resolution due, in large part, to spatial separation between stimulating electrode arrays and primary neural receptors. In this work, a combination of physical and chemical micropatterns, formed on acrylate polymers, are used to
Violaine Augustin et al.
Electrophoresis, 29(18), 3875-3886 (2008-10-14)
A polyacrylate-based monolithic column bearing cationic functionalities and designed for capillary electrochromatography (CEC) has been prepared via photopolymerization of a mixture of hexyl acrylate, butanediol diacrylate, 2-(acryloyloxy) ethyltrimethyl ammonium chloride (monomers), azobisisobutyronitrile (photoinitiator), acetonitrile, phosphate buffer, and ethanol (porogens). The
Emilia Stelmach et al.
Talanta, 163, 17-23 (2016-11-26)
Analytical benefits related to application of copolymeric microspheres containing different number of carboxylic acid mers have been studied on example of acrylate copolymers. These structures can be used as a reagent in heterogeneous pH titration, benefiting from different number of
Amrita Sarkar et al.
Soft matter, 15(26), 5193-5203 (2019-06-18)
The combination of precision control with wide tunability remains a challenge for the fabrication of porous nanomaterials suitable for studies of nanostructure-behavior relationships. Polymer micelle templates broadly enable porous materials, however micelle equilibration hampers independent pore and wall size control.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service