Skip to Content
Merck
All Photos(1)

Key Documents

407682

Sigma-Aldrich

4-(Trifluoromethyl)pyrimidine-2-thiol

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H3F3N2S
CAS Number:
Molecular Weight:
180.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

solid

mp

164-166 °C (lit.)

functional group

fluoro

SMILES string

FC(F)(F)c1ccnc(S)n1

InChI

1S/C5H3F3N2S/c6-5(7,8)3-1-2-9-4(11)10-3/h1-2H,(H,9,10,11)

InChI key

HDGJASCXBNRLGA-UHFFFAOYSA-N

General description

4-(Trifluoromethyl)pyrimidine-2-thiol is a heteroaromatic thioamide. Separation and quantification of a mixture of heteroaromatic thioamides (pyridine-2-thione, 5-trifluoromethylpyridine-2-thione, pyrimidine-2-thiol, 4-trifluoromethylpyrimidine-2-thiol) by capillary zone electrophoresis has been reported.

Application

4-(Trifluoromethyl)pyrimidine-2-thiol may be used in the preparation of [{Pt(bzq)(μ-L)}2] [Hbzq = benzo[h]quinolone, HL = 4-(trifluoromethyl)pyrimidine-2-thiol], a novel half-lantern divalent complex of platinum.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrophoretic determination of thioamides, pyridine and pyrimidine derivatives.
Chenov'yants MS, et al.
Journal of Analytical Chemistry, 68(6), 532-535 (2013)
Stayin' Alive-Organoplatinum Complexes.
Klein A.
Inorganics , 3(2), 155-159 (2015)
Half-Lantern Pt (II) and Pt (III) Complexes. New Cyclometalated Platinum Derivatives.
Sicilia V, et al.
Inorganics , 2(3), 508-523 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service