Skip to Content
Merck
All Photos(1)

Key Documents

407577

Sigma-Aldrich

Trimethylolpropane propoxylate triacrylate

average Mn ~644

Synonym(s):

Propoxylated trimethylolpropane triacrylate

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$148.00

$148.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$148.00

About This Item

Linear Formula:
[H2C=CHCO2(C3H6O)nCH2]3CC2H5
CAS Number:
Molecular Weight:
644.79
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$148.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

liquid

mol wt

average Mn ~644

contains

500 ppm monomethyl ether hydroquinone as inhibitor

refractive index

n20/D 1.46

density

1.051 g/mL at 25 °C

SMILES string

CCC(COCCCOC(=O)C=C)(COCCCOC(=O)C=C)COCCCOC(=O)C=C

InChI key

NCNNNERURUGJAB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service