Skip to Content
Merck
All Photos(2)

Key Documents

407445

Sigma-Aldrich

4-(4-Bromophenyl)-4-piperidinol

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$126.00

$126.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$126.00

About This Item

Empirical Formula (Hill Notation):
C11H14BrNO
CAS Number:
Molecular Weight:
256.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$126.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

mp

167-170 °C (lit.)

functional group

bromo
hydroxyl

SMILES string

OC1(CCNCC1)c2ccc(Br)cc2

InChI

1S/C11H14BrNO/c12-10-3-1-9(2-4-10)11(14)5-7-13-8-6-11/h1-4,13-14H,5-8H2

InChI key

QNLXJYQUWCNYBH-UHFFFAOYSA-N

General description

4-(4-Bromophenyl)-4-piperidinol is reported to show potential antioxidant activity. Analgesic effects of 4-(4-bromophenyl)-4-piperidinol in mice has been studied using a tail immersion assay.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zafar Saeed Saify et al.
Archives of pharmacal research, 35(11), 1953-1959 (2012-12-06)
Piperidine derivatives are known to exhibit analgesic activities and are likely to possess the ability to block the effects of prostaglandins through inhibition of downstream signaling pathways. The present study investigated the activity of five derivatives (PD2-6) of 4-(4'-bromophenyl)-4-piperidinol (PD1)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service