Skip to Content
Merck
All Photos(1)

Key Documents

404756

Sigma-Aldrich

N-Benzylphthalimide

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11NO2
CAS Number:
Molecular Weight:
237.25
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

114-116 °C (lit.)

SMILES string

O=C1N(Cc2ccccc2)C(=O)c3ccccc13

InChI

1S/C15H11NO2/c17-14-12-8-4-5-9-13(12)15(18)16(14)10-11-6-2-1-3-7-11/h1-9H,10H2

InChI key

WITXFYCLPDFRNM-UHFFFAOYSA-N

General description

N-Benzylphthalimide (NBPT), also known as 2-benzylisoindoline-1,3-dione, is an N-substituted phthalimide. It has been prepared by reacting phthalic anhydride with benzyl amine in glacial acetic acid. Vibrational spectra of NBPT has been recorded and assigned.[1] NBPT is a roof-shaped molecule with a planar cyclic imide and a phenyl ring connected by a methylene group.[2] Crystal structure of N-benzylphthalimide has parallel layers of phthalimides stack along the a axis.[2]

Application

N-Benzylphthalimide may be used in the following syntheses:
  • 2-benzyl-1,1,3,3-tetraphenylisoindoline[3]
  • tailor-made highly fluorous porphyrin derivatives[4]
  • N-benzylisoindole[5]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Impact of molecular size on electron spin relaxation rates of nitroxyl radicals in glassy solvents between 100 and 300 K.
Sato HIDEO, et al.
Molecular Physics, 105(15-16), 2137-2151 (2007)
Highly fluorous porphyrins as model compounds for molecule interferometry.
Tuxen J, et al.
European Journal of Organic Chemistry, 25, 4823-4833 (2011)
N-benzylphthalimide.
Warzecha K-D, et al.
Acta Crystallographica Section E, Structure Reports Online, 62(6), 2367-2368 (2006)
Vibrational assignment of N-benzylphthalimide and 15N-benzylphthalimide.
Kolev T and Juchnovski I.
Journal of Molecular Structure, 349, 377-380 (1995)
The direct conversion of phthalimides to isoindoles.
Garmaise DL and Ryan A.
Journal of Heterocyclic Chemistry, 7(2), 413-413 (1970)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service