Skip to Content
Merck
All Photos(2)

Key Documents

403644

Sigma-Aldrich

N-(5-Chloro-2-pyridyl)bis(trifluoromethanesulfonimide)

96%

Synonym(s):

2-[N,N-Bis(trifluoromethylsulfonyl)amino]-5-chloropyridine, N,N-Bis(trifluoromethylsulfonyl)-5-chloro-2-pyridylamine, Comins’ Reagent

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$37.10
5 G
$164.00

$37.10

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$37.10
5 G
$164.00

About This Item

Empirical Formula (Hill Notation):
C7H3ClF6N2O4S2
CAS Number:
Molecular Weight:
392.68
Beilstein:
5833971
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$37.10

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Assay

96%

form

solid

mp

45-47 °C (lit.)

functional group

chloro
fluoro

storage temp.

2-8°C

SMILES string

FC(F)(F)S(=O)(=O)N(c1ccc(Cl)cn1)S(=O)(=O)C(F)(F)F

InChI

1S/C7H3ClF6N2O4S2/c8-4-1-2-5(15-3-4)16(21(17,18)6(9,10)11)22(19,20)7(12,13)14/h1-3H

InChI key

TUFGVZMNGTYAQD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Provides good yields of vinyl triflates from the corresponding ketone enolates or dienolates.[1][2][3]
Reactant for:
Suzuki-Miyaura cross coupling
Synthesis of nicotinic acetylcholine receptor-selective ligands
Enantioselective desymmetrizing palladium catalyzed carbonylation reactions
Synthesis of high affinity niacin receptor GPR109A agonists
Preparation of heteroaromatics

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 44, 9185-9185 (2003)
Synthesis, 4, 588-588 (2000)
Tetrahedron Letters, 33, 6299-6299 (1992)
Ian Paterson et al.
Angewandte Chemie (International ed. in English), 53(10), 2692-2695 (2014-02-01)
Leiodermatolide is an antimitotic macrolide isolated from the marine sponge Leiodermatium sp. whose potentially novel tubulin-targeting mechanism of action makes it an exciting lead for anticancer drug discovery. In pursuit of a sustainable supply, we report a highly stereocontrolled total

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service