Skip to Content
Merck
All Photos(1)

Key Documents

403253

Sigma-Aldrich

Zirconium-nickel alloy

powder, −325 mesh

Synonym(s):

Ni5Zr2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ni5Zr2
CAS Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Pricing and availability is not currently available.

form

powder

composition

Ni, 70%
Zr, 30%

particle size

−325 mesh

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Eye Dam. 1 - Flam. Sol. 1 - Skin Sens. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service