Skip to Content
Merck
All Photos(1)

Key Documents

402435

Sigma-Aldrich

2-Amino-7-bromo-5-oxo-5H-[1]benzopyrano[2,3-b]pyridine-3-carbonitrile

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H6BrN3O2
CAS Number:
Molecular Weight:
316.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

>300 °C (lit.)

functional group

bromo
ketone
nitrile

SMILES string

Nc1nc2Oc3ccc(Br)cc3C(=O)c2cc1C#N

InChI

1S/C13H6BrN3O2/c14-7-1-2-10-8(4-7)11(18)9-3-6(5-15)12(16)17-13(9)19-10/h1-4H,(H2,16,17)

InChI key

UFLWUJOAQJLVMJ-UHFFFAOYSA-N

General description

2-Amino-7-bromo-5-oxo-5H-[1]benzopyrano[2,3-b]pyridine-3-carbonitrile is an halogenated heterocycle.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tyler S Beyett et al.
Bioorganic & medicinal chemistry, 26(20), 5443-5461 (2018-10-03)
The non-canonical IκB kinases TANK-binding kinase 1 (TBK1) and inhibitor of nuclear factor kappa-B kinase ε (IKKε) play a key role in insulin-independent pathways that promote energy storage and block adaptive energy expenditure during obesity. Utilizing docking calculations and the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service