Skip to Content
Merck
All Photos(1)

Key Documents

401730

Sigma-Aldrich

Methyl 10-chloro-10-oxodecanoate

97%

Synonym(s):

Methyl sebacoyl chloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$222.00

$222.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$222.00

About This Item

Linear Formula:
ClCO(CH2)8CO2CH3
CAS Number:
Molecular Weight:
234.72
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$222.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

form

liquid

refractive index

n20/D 1.453 (lit.)

bp

159-160 °C/9 mmHg (lit.)

density

1.052 g/mL at 25 °C (lit.)

functional group

acyl chloride
ester

SMILES string

COC(=O)CCCCCCCCC(Cl)=O

InChI

1S/C11H19ClO3/c1-15-11(14)9-7-5-3-2-4-6-8-10(12)13/h2-9H2,1H3

InChI key

ZUUUAAZQEMFVSN-UHFFFAOYSA-N

General description

Methyl 10-chloro-10-oxodecanoate (Methyl sebacoyl chloride, 10-Oxo-10-chlorodecanoic acid methyl ester) is a monomethyl ester of sebacinic acid.

Application

Methyl 10-chloro-10-oxodecanoate may be used in the preparation of the following:
  • methyl 10-[2-methoxy-4-(3-trifluoromethyl-3H-diazirin-3-yl)phenyl]-10-oxodecanoate[1]
  • fluorescein-derivatives of Necrostatin-3[2]
  • methyl 10-oxo-10-(Pyren-3-yl)decanoate[3]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jenny L Maki et al.
Analytical biochemistry, 427(2), 164-174 (2012-06-05)
Necrotic cell death is prevalent in many different pathological disease states and in traumatic injury. Necroptosis is a form of necrosis that stems from specific signaling pathways, with the key regulator being receptor interacting protein 1 (RIP1), a serine/threonine kinase.
Effective synthesis of a carbon-linked diazirinyl fatty acid derivative via reduction of the carbonyl group to methylene with triethylsilane and trifluoroacetic acid.
Hashimoto M, et al.
Heterocycles, 59(1), 395-398 (2003)
Wallace Thompson et al.
Methods in molecular biology (Clifton, N.J.), 861, 149-158 (2012-03-20)
Attempts to characterize, quantify, and/or modulate the activity of the secreted phospholipase A(2) family of enzymes result from the diversity of physiological roles for which these enzymes have been implicated. The 1-palmitoyl-2-(10-pyrenedecanoyl)-phosphatidylglycerol (pyrenePG)-based fluorometric assay is a sensitive and readily

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service