Skip to Content
Merck
All Photos(2)

Key Documents

401277

Sigma-Aldrich

Cesium oxalate

≥99.9% trace metals basis

Synonym(s):

Oxalic acid cesium salt

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$76.40

$76.40


Estimated to ship onMay 29, 2025


Request a Bulk Order

Select a Size

Change View
5 G
$76.40

About This Item

Linear Formula:
(COOCs)2
CAS Number:
Molecular Weight:
353.83
Beilstein:
3709030
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder

$76.40


Estimated to ship onMay 29, 2025


Request a Bulk Order

grade

for analytical purposes

Assay

≥99.9% trace metals basis

form

powder

impurities

≤1000.0 ppm Trace Metal Analysis

SMILES string

[Cs+].[Cs+].[O-]C(=O)C([O-])=O

InChI

1S/C2H2O4.2Cs/c3-1(4)2(5)6;;/h(H,3,4)(H,5,6);;/q;2*+1/p-2

InChI key

HEQUOWMMDQTGCX-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Egor Evlyukhin et al.
Physical chemistry chemical physics : PCCP, 20(28), 18949-18956 (2018-06-06)
X-ray induced damage has been known for decades and has largely been viewed as a tremendous nuisance. We, on the other hand, harness the highly ionizing and penetrating properties of hard X-rays to initiate novel decomposition and synthetic chemistry. Here

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service