Skip to Content
Merck
All Photos(2)

Key Documents

401188

Sigma-Aldrich

Antimony(III) iodide

98%

Synonym(s):

Antimony triiodide

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$71.30

$71.30

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$71.30

About This Item

Linear Formula:
SbI3
CAS Number:
Molecular Weight:
502.47
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
crystalline
solubility:
acetone: slightly soluble(lit.)
ethanol: slightly soluble(lit.)

$71.30

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

for analytical purposes

Quality Level

Assay

98%

form

crystalline

reaction suitability

reagent type: catalyst
core: antimony

bp

401 °C (lit.)

mp

170 °C (lit.)

solubility

acetone: slightly soluble(lit.)
ethanol: slightly soluble(lit.)

density

4.92 g/mL at 25 °C (lit.)

SMILES string

I[Sb](I)I

InChI

1S/3HI.Sb/h3*1H;/q;;;+3/p-3

InChI key

KWQLUUQBTAXYCB-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katayoon Tabatabaei et al.
ACS nano, 15(1), 1685-1700 (2021-01-13)
Doped and alloyed germanium nanocrystals (Ge NCs) are potential candidates for a variety of applications such as photovoltaics and near IR detectors. Recently, bismuth (Bi) as an n-type group 15 element was shown to be successfully and kinetically doped into
A Gągor et al.
Dalton transactions (Cambridge, England : 2003), 46(47), 16605-16614 (2017-11-23)
The easy to prepare and stable in air (2-methylimidazolium) tetraiodoantimoniate(iii) single-crystals with optical band gap of 2.17(1) eV at room temperature have been synthesized. The crystal structure features one-dimensional [SbI
Jian Zhang et al.
ACS nano, 11(9), 9294-9302 (2017-09-08)
Colloidal quantum dots (QDs) of lead halide perovskite have recently received great attention owing to their remarkable performances in optoelectronic applications. However, their wide applications are hindered from toxic lead element, which is not environment- and consumer-friendly. Herein, we utilized
O S Urgut et al.
Materials science & engineering. C, Materials for biological applications, 58, 396-408 (2015-10-20)
Antimony(III) halide complexes of the formulae {[SbBr(Me2DTC)2]n} (1), {[SbI(Me2DTC)2]n} (2) and {[(Me2DTC)2Sb(μ2-I)Sb(Me2DTC)2](+).I3(-)} (3) (Me2DTC = dimethyldithiocarbomate) were synthesized from SbX3, (X = Br or I) and tetramethylthiuram monosulfide (Me4tms) or tetramethylthiuram disulfide (Me4tds). The complexes were characterized by melting point

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service