All Photos(1)
About This Item
Linear Formula:
TaBr5
CAS Number:
Molecular Weight:
580.47
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Assay
98%
form
powder
particle size
-8 mesh
mp
240 °C (lit.)
density
4.989 g/mL at 25 °C (lit.)
SMILES string
Br[Ta](Br)(Br)(Br)Br
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Ta6Br(2+)12, a tool for phase determination of large biological assemblies by X-ray crystallography.
J Knäblein et al.
Journal of molecular biology, 270(1), 1-7 (1997-07-04)
The title compound Ta6Br(2+)12 is of interest for the analysis of biological structures as a heavy-metal derivative with great potential for the structure determination of large protein systems. In macromolecular crystallography the phases of the measured structure factor amplitudes have
L Ditzel et al.
Biological chemistry, 378(3-4), 239-247 (1997-03-01)
The novel proteolytic mechanism of the 20S proteasome from T. acidophilum has been investigated by X-ray crystallography using small-molecule inhibitors and substrate analogues. The 20S proteasome degrades unfolded substrates into small peptides of a defined length. Calpain inhibitor II, chymostatin
F X Gomis-Rüth et al.
Acta crystallographica. Section D, Biological crystallography, 57(Pt 6), 800-805 (2001-05-26)
The structure of the conjugative coupling protein TrwBDeltaN70 from Escherichia coli plasmid R388 was solved using two crystal forms. This large multimeric membrane protein of 437 residues per monomer is involved in cell-to-cell single-strand DNA transfer. Diffraction data to 2.4
Sankaran Banumathi et al.
Acta crystallographica. Section D, Biological crystallography, 59(Pt 3), 492-498 (2003-02-22)
The Ta(6)Br(12)(2+) cluster compound is known to be a powerful reagent for derivatization of crystals of large macromolecules at low resolution. The cluster is a regular octahedron of six Ta atoms with 12 bridging Br atoms at the edges of
T Neuefeind et al.
Biological chemistry, 378(3-4), 219-221 (1997-03-01)
The title compound Ta6Br12(2+) is of interest for the analysis of biological structures as a heavy metal derivative with great potential for the structure determination of large protein systems. In macromolecular crystallography the phases of the measured structure factor amplitudes
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service