Skip to Content
Merck
All Photos(1)

Key Documents

399817

Sigma-Aldrich

Titanium(III) fluoride

Synonym(s):

Titanium trifluoride

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$82.40

$82.40


Estimated to ship onMay 25, 2025


Request a Bulk Order

Select a Size

Change View
5 G
$82.40

About This Item

Linear Formula:
TiF3
CAS Number:
Molecular Weight:
104.86
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$82.40


Estimated to ship onMay 25, 2025


Request a Bulk Order

form

powder

reaction suitability

core: titanium
reagent type: catalyst

density

3.4 g/mL at 25 °C (lit.)

SMILES string

F[Ti](F)F

InChI

1S/3FH.Ti/h3*1H;/q;;;+3/p-3

InChI key

NLPMQGKZYAYAFE-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Titanium dioxide applications: Semiconducting material characteristics and diverse functionalities.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service