Skip to Content
Merck
All Photos(2)

Key Documents

396591

Sigma-Aldrich

4-(Methylthio)phenylacetic acid

99%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$76.20

$76.20


Estimated to ship onMay 25, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$76.20

About This Item

Linear Formula:
CH3SC6H4CH2CO2H
CAS Number:
Molecular Weight:
182.24
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$76.20


Estimated to ship onMay 25, 2025


Request a Bulk Order

Assay

99%

mp

97-98 °C (lit.)

functional group

carboxylic acid
thioether

SMILES string

CSc1ccc(CC(O)=O)cc1

InChI

1S/C9H10O2S/c1-12-8-4-2-7(3-5-8)6-9(10)11/h2-5H,6H2,1H3,(H,10,11)

InChI key

AHMLFHMRRBJCRM-UHFFFAOYSA-N

General description

TiO2 photocatalytic one-electron oxidation of 4-(methylthio)phenylacetic acid has been reported by time-resolved diffuse reflectance spectroscopy.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Influences of adsorption on TiO2 photocatalytic one-electron oxidation of aromatic sulfides studied by time-resolved diffuse reflectance spectroscopy.
Tachikawa T, et al.
The Journal of Physical Chemistry B, 108(19), 5859-5866 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service