Skip to Content
Merck
All Photos(1)

Key Documents

396265

Sigma-Aldrich

Bis(2-carboxyethylgermanium(IV) sesquioxide)

99%

Synonym(s):

GE 132

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$106.00

$106.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$106.00

About This Item

Linear Formula:
O[Ge(=O)CH2CH2CO2H]2
CAS Number:
Molecular Weight:
339.42
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$106.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

99%

form

solid

functional group

carboxylic acid

SMILES string

OC(=O)CC[Ge](=O)O[Ge](=O)CCC(O)=O

InChI

1S/C6H10Ge2O7/c9-5(10)1-3-7(13)15-8(14)4-2-6(11)12/h1-4H2,(H,9,10)(H,11,12)

InChI key

XEABSBMNTNXEJM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bis(2-carboxyethylgermanium(IV) sesquioxide), also known as repagermanium, is a water-soluble organogermanium compound that exhibits immunostimulatory properties. It is also used as an ingredient in food and cosmetics.[1]

Application

Bis(2-carboxyethylgermanium(IV) sesquioxide) can be used as a precursor in the preparation of photoluminescent carbon dots, applicable as fluorescent probes in bioimaging applications.[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yun Huan Yuan et al.
Nanoscale, 7(40), 16841-16847 (2015-09-26)
Carbon dots doped with germanium (GeCDs) were firstly prepared by a new simple 15 min carbonation synthesis route, exhibiting excitation-independent photoluminescence (PL), which could avoid autofluorescence in bioimaging applications. The as-prepared GeCDs have low cell toxicity, good biocompatibility, high intracellular
Yukinori Tamura et al.
Arteriosclerosis, thrombosis, and vascular biology, 28(12), 2195-2201 (2008-09-27)
Recently, adipose tissue inflammation induced by macrophage infiltration through MCP-1/C-C chemokine receptor-2 (CCR2) pathway is considered to play a role in the development of visceral obesity and insulin resistance. In the present study, to further examine the role of CCR2
Petra Mulder et al.
PloS one, 12(1), e0169740-e0169740 (2017-01-12)
Obese patients with chronic inflammation in white adipose tissue (WAT) have an increased risk of developing non-alcoholic steatohepatitis (NASH). The C-C chemokine receptor-2 (CCR2) has a crucial role in the recruitment of immune cells to WAT and liver, thereby promoting
Bonnie J Kaplan et al.
Journal of alternative and complementary medicine (New York, N.Y.), 10(2), 337-344 (2004-05-29)
This paper reviews the history, chemistry, safety, toxicity, and anticancer effects of the organogermanium compound bis (2-carboxyethylgermanium) sesquioxide (CEGS). A companion review follows, discussing the inaccuracies in the scientific record that have prematurely terminated research on clinical uses of CEGS.
Chisato Hirayama et al.
Journal of gastroenterology, 38(6), 525-532 (2003-06-26)
Although antiviral agents have been adopted for the management of chronic hepatitis B, they have only limited efficacy because of the underlying impaired immune status. Propagermanium, a hydrophilic polymer of 3-oxygermyl propionate, has been reported to have potent immune modulatory

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service