Skip to Content
Merck
All Photos(1)

Key Documents

394017

Sigma-Aldrich

Zinc trifluoroacetate hydrate

Synonym(s):

Trifluoroacetic acid zinc salt

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$58.90

$58.90

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$58.90

About This Item

Linear Formula:
Zn(CF3COO)2 · xH2O
CAS Number:
Molecular Weight:
291.42 (anhydrous basis)
Beilstein:
3719156
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

$58.90

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

composition

Zn, ≤21.8% EDTA titration

reaction suitability

reagent type: catalyst

SMILES string

FC(F)(F)C(=O)O[Zn]OC(=O)C(F)(F)F

InChI

1S/2C2HF3O2.Zn/c2*3-2(4,5)1(6)7;/h2*(H,6,7);/q;;+2/p-2

InChI key

VCQWRGCXUWPSGY-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Few Monolayer Atomic Layer Deposition (ALD) on Surfaces and Interfaces for Energy Applications

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service