Skip to Content
Merck
All Photos(2)

Documents

392081

Sigma-Aldrich

Tris(2,4,6-trimethoxyphenyl)phosphine

Synonym(s):

TTMPP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3O)3C6H2]3P
CAS Number:
Molecular Weight:
532.52
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.22

form

solid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

mp

155-160 °C (lit.)

functional group

phosphine

SMILES string

COc1cc(OC)c(P(c2c(OC)cc(OC)cc2OC)c3c(OC)cc(OC)cc3OC)c(OC)c1

InChI

1S/C27H33O9P/c1-28-16-10-19(31-4)25(20(11-16)32-5)37(26-21(33-6)12-17(29-2)13-22(26)34-7)27-23(35-8)14-18(30-3)15-24(27)36-9/h10-15H,1-9H3

InChI key

JQKHNBQZGUKYPX-UHFFFAOYSA-N

Related Categories

Application

Highly basic arylphosphine.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of the Chemical Society. Chemical Communications, 482-482 (1984)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service