Skip to Content
Merck
All Photos(1)

Key Documents

391115

Sigma-Aldrich

N-(Phenylthio)phthalimide

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H9NO2S
CAS Number:
Molecular Weight:
255.29
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

mp

160-163 °C (lit.)

solubility

DMSO: soluble 25 mg/mL, clear, yellow

SMILES string

O=C1N(Sc2ccccc2)C(=O)c3ccccc13

InChI

1S/C14H9NO2S/c16-13-11-8-4-5-9-12(11)14(17)15(13)18-10-6-2-1-3-7-10/h1-9H

InChI key

NMHKBABHRKQHOL-UHFFFAOYSA-N

General description

N-(Phenylthio)phthalimide is an N-substituted phthalimide. It is used as a sulphenylating agent. The synthesis of N-(phenylthio)phthalimide has been reported.

Application

N-(Phenylthio)phthalimide is suitable reagent used in the synthesis of phenyl (3-trimethoxysilylpropyl)-disulfide. It may be used in the sulfenylation of ylides and β−keto esters.
N-(Phenylthio)phthalimide may be used as sulfenylating agent in the synthesis of the following:
  • α-phenylthio-ketones and α-phenylthio-aldehydes from aldehydes and ketones
  • 3-sulfenyl indoles from indoles
  • optically active 3-phenylthiooxindoles from unprotected 3-substituted oxindoles
  • N−alkyl-β-phenylsulfenyl-2-acetylbenzimidazole from N−alkyl−2−acetylbenzimidazole
  • N−phenylsulfenyl ketimines from oximes or nitro compounds

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Direct, organocatalytic α-sulfenylation of aldehydes and ketones.
Wang W, et al.
Tetrahedron Letters, 45(44), 8229-8231 (2004)
A new facile sulfenylation of 2-acetylbenzimidazoles.
Kumar PP, et al.
Phosphorus, Sulfur, and Silicon and the Related Elements, 189(12), 1775-1779 (2014)
The use of anhydrous CeCl3 as a catalyst for the synthesis of 3-sulfenyl indoles.
Silveira CC, et al.
Tetrahedron Letters, 51(15), 2014-2016 (2010)
Jordi Burés et al.
Organic letters, 9(22), 4635-4638 (2007-10-09)
As N-sulfenyl imines (e.g., RR'C=N-SAr) can be readily transformed to their N-sulfinyl imines (RR'C=N-SOAr), N-sulfonyl imines (RR'C=N-SO2Ar), and N-sulfonyl oxaziridines, the very mild procedure developed to convert ketoximes and secondary nitro derivatives to N-arenesulfenyl ketimines constitutes a new and efficient
On the involvement of lipoic acid in. alpha.-keto acid dehydrogenase complexes.
Rastetter WH, et al.
Journal of the American Chemical Society, 101(10), 2752-2753 (1979)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service