Skip to Content
Merck
All Photos(1)

Key Documents

390690

Sigma-Aldrich

(2-Hydroxypropyl)-α-cyclodextrin

average Mw ~1,180

Synonym(s):

α‐HPCD, 2OHpαCD, 2‐hydroxypropyl‐α‐CD, HPαCD, HP-α-CD

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$80.80
25 G
$261.00

$80.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$80.80
25 G
$261.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.22

$80.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

optical activity

[α]20/D +125°, c = 1 in H2O

mol wt

average Mw ~1,180

extent of labeling

0.6 molar substitution

mp

245 °C (dec.) (lit.)

SMILES string

O1[C@H]2O[C@@H]([C@@H](O[C@H]3O[C@@H]([C@@H](O[C@H]4O[C@@H]([C@@H](O[C@H]5O[C@@H]([C@@H](O[C@H]6O[C@@H]([C@@H](O[C@H]7O[C@@H]([C@@H]1[C@H]([C@@H]7O)O)COCC(O)C)[C@H]([C@@H]6O)O)COCC(O)C)[C@H]([C@@H]5O)O)COCC(O)C)[C@H]([C@@H]4O)O)COCC(O)C)[C@H]([C@@H]3O)O)C

InChI

1S/C54H96O36/c1-19(55)7-73-13-25-43-31(61)37(67)49(79-25)86-44-26(14-74-8-20(2)56)81-51(39(69)33(44)63)88-46-28(16-76-10-22(4)58)83-53(41(71)35(46)65)90-48-30(18-78-12-24(6)60)84-54(42(72)36(48)66)89-47-29(17-77-11-23(5)59)82-52(40(70)34(47)64)87-45-27(15

InChI key

PMPYGZCSMCXHTA-WABPNHHRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(2-Hydroxypropyl)-α-cyclodextrin is a hydroxyalkyl derivative of cyclodextrin, which is generally used as a substitute of α-cyclodextrin for potential application in food, cosmetics and pharmaceutical industries. It may be used as a parenteral drug carrier on account of its attractive characteristics such as: a greater extent of solubility in water, high dissolution capacity for weakly soluble analytes, and low toxicity.[1][2]

Application

  • (2-Hydroxypropyl)-α-cyclodextrin can be used as a chiral selector to resolve tryptophan racemate by capillary electrophoresis.[3]
  • It can be used to synthesize biodegradable polyrotaxanes for fabricating blood-contacting devices.[4]
  • Co-polymer based on (2-hydroxypropyl)-α-cyclodextrin cross-linked with polyethylenimine can be used as a gene delivery vector.[5]
  • It can also be used as stabilizer for antibacterial iodine formulations, as it forms a stable inclusion complex with iodine and improves aqueous solubility.[6]

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

2-Hydroxypropylated cyclodextrins as a sustained-release carrier for fragrance materials
TANAKA M, et al.
Chemical & Pharmaceutical Bulletin, 44(2), 416-420 (1996)
Interaction of iodine with 2-hydroxypropyl-α-cyclodextrin and its bactericidal activity.
Tomono K, et al.
Drug Development and Industrial Pharmacy, 28(10), 1303-1309 (2002)
Effect of biodegradable polyrotaxanes on platelet activation.
Yui N, et al.
Bioconjugate Chemistry, 9(1), 118-125 (1998)
A novel co-polymer based on hydroxypropyl α-cyclodextrin conjugated to low molecular weight polyethylenimine as an in vitro gene delivery vector.
Huang H, et al.
International Journal of Molecular Sciences, 9(11), 2278-2289 (2008)
Recognition mechanism of d-and l-tryptophan enantiomers using 2-hydroxypropyl-α-or β-cyclodextrins as chiral selectors.
Malta LFB, et al.
Tetrahedron Asymmetry, 19(10), 1182-1188 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service