Skip to Content
Merck
All Photos(1)

Documents

390321

Sigma-Aldrich

Trifluoroacetaldehyde dimethyl acetal

99%

Synonym(s):

1,1,1-Trifluoro-2,2-dimethoxyethane, 2,2-Dimethoxy-1,1,1-trifluoroethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3CH(OCH3)2
CAS Number:
Molecular Weight:
144.09
Beilstein:
2070944
MDL number:
UNSPSC Code:
12352103

Assay

99%

refractive index

n20/D 1.320 (lit.)

bp

78-79 °C (lit.)

density

1.2 g/mL at 25 °C (lit.)

SMILES string

COC(OC)C(F)(F)F

Looking for similar products? Visit Product Comparison Guide

Application

Alternative to gaseous unstable trifluoroacetaldehyde used to prepare α-trifluoromethylated alcohols for antifungals, antitumor, and chemotherapeutic agents.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 32, 3823-3823 (1991)
Tetrahedron Asymmetry, 2, 235-235 (1991)
Tetrahedron Letters, 33, 1351-1351 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service