Skip to Content
Merck
All Photos(1)

Documents

386626

Sigma-Aldrich

Aluminum 1,4,8,11,15,18,22,25-octabutoxy-29H,31H-phthalocyanine triethylsiloxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C70H95AlN8O9Si
CAS Number:
Molecular Weight:
1247.62
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

mp

100 °C (dec.) (lit.)

λmax

766 nm

SMILES string

CCCCOc1ccc(OCCCC)c2c3nc(nc4n5[Al](O[Si](CC)(CC)CC)n6c(n3)c7c(OCCCC)ccc(OCCCC)c7c6nc8nc(nc5c9c(OCCCC)ccc(OCCCC)c49)c%10c(OCCCC)ccc(OCCCC)c8%10)c12


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service