Skip to Content
Merck
All Photos(1)

Key Documents

385409

Sigma-Aldrich

4-Bromo-3-methylanisole

98%

Synonym(s):

2-Bromo-5-methoxytoluene

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
$184.00

$184.00


Estimated to ship onApril 05, 2025


Request a Bulk Order

Select a Size

Change View
25 ML
$184.00

About This Item

Linear Formula:
BrC6H3(CH3)OCH3
CAS Number:
Molecular Weight:
201.06
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$184.00


Estimated to ship onApril 05, 2025


Request a Bulk Order

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.561 (lit.)

bp

111-112 °C/15 mmHg (lit.)

solubility

water: soluble (Partially miscible)(lit.)

density

1.424 g/mL at 25 °C (lit.)

functional group

bromo

SMILES string

COc1ccc(Br)c(C)c1

InChI

1S/C8H9BrO/c1-6-5-7(10-2)3-4-8(6)9/h3-5H,1-2H3

InChI key

BLZNSXFQRKVFRP-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of cyclodextrins on electrophilic aromatic bromination in aqueous solution.
<BIG>Dumanski et al.</BIG>
Australian Journal of Chemistry, 56(11), 1107-1111 (2003)
The synthesis of (?)-aplysin and (?)-debromoaplysin.
<BIG>Yamada et al. </BIG>
Chemical Communications (Cambridge, England), 22, 1432-1432 (1968)
Discovery of orally active nonpeptide vitronectin receptor antagonists based on a 2-benzazepine Gly-Asp mimetic.
W H Miller et al.
Journal of medicinal chemistry, 43(1), 22-26 (2000-01-14)
A concise synthesis of the naphthoic acid component of neocarzinostatin chromophore featuring a new photocyclization reaction.
Myers et al.
Tetrahedron Letters, 37.5, 587-590 (1996)
Lucas R Moore et al.
Organic letters, 6(2), 225-228 (2004-01-16)
[reaction: see text] Sterically demanding, sulfonated arylphosphines TXPTS and TMAPTS have been applied to the aqueous-phase Heck and Suzuki coupling of aryl bromides. TXPTS provides good yields of Heck coupling products from aryl bromides at 80 degrees C, while both

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service