Skip to Content
Merck
All Photos(1)

Key Documents

383295

Sigma-Aldrich

Barium ferrite

−325 mesh, 98%

Synonym(s):

Barium ferrite

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Linear Formula:
BaFe12O19
CAS Number:
Molecular Weight:
1111.46
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

98%

form

powder

particle size

−325 mesh

SMILES string

O=[Ba].O=[Fe]O[Fe]=O.O=[Fe]O[Fe]=O.O=[Fe]O[Fe]=O.O=[Fe]O[Fe]=O.O=[Fe]O[Fe]=O.O=[Fe]O[Fe]=O

InChI

1S/Ba.12Fe.19O

InChI key

AJCDFVKYMIUXCR-UHFFFAOYSA-N

Specificity

May contain varying amounts of oxide.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - STOT RE 2 - STOT SE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A M Granov et al.
Vestnik rentgenologii i radiologii, (5)(5), 53-54 (1993-09-01)
Various types of x-ray endovascular embolization were carried out in 256 patients with renal parenchymatous carcinoma over the period of 1979 to 1991. Iodolipol chemoembolization with dioxadet, a fat-soluble preparation, significantly improved three- and five-year cumulative survival of operable patients
[Specialized outpatient care in infectious diseases].
E N Antakova et al.
Vrachebnoe delo, (7)(7), 113-115 (1982-07-01)
Elif Tümay Özer et al.
Journal of hazardous materials, 229-230, 20-28 (2012-07-04)
The barium hexaferrite (BaFe(12)O(19)) containing magnetic poly(ethylene glycol dimethacrylate-vinyl pyridine), (mag-poly(EGDMA-VP)) beads (average diameter=53-212 μm) were synthesized and characterized. Their use as an adsorbent in the removal of diethyl phthalate (DEP) from an aqueous solution was investigated. The mag-poly(EGDMA-VP) beads

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service