Skip to Content
Merck
All Photos(1)

Key Documents

378135

Sigma-Aldrich

Beryllium

powder, −325 mesh, ≥99%

Synonym(s):

Glucinium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Be
CAS Number:
Molecular Weight:
9.01
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

≥99%

form

powder

autoignition temp.

1198 °F

reaction suitability

reagent type: Lewis acid
core: beryllium

reagent type: catalyst

resistivity

4.46 μΩ-cm, 20°C

particle size

−325 mesh

bp

2970 °C (lit.)

mp

1278 °C (lit.)

density

1.85 g/mL at 25 °C (lit.)

SMILES string

[Be]

InChI

1S/Be

InChI key

ATBAMAFKBVZNFJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Starting material for novel sterically encumbered arylberyllium compounds.[1]

Reagent for synthesis of:
  • Beryllium phthalocyanine and its derivatives[2]
  • Catalyst with cerium oxide for oxidative coupling of methane[3]
  • Organoberyllium compounds[4]

Reagent for:
  • Metal mediated dechlorination[5]
  • Stereoselective Horner-Wadsworth-Emmons reactions[6]

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Carc. 1B Inhalation - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

I. I. Lapkin and S. V. Sinani,
Zh. Obshch. Khim., 57, 372-375 (1987)
K. Otsuka, et al.,
Chemistry Letters (Jpn), 9, 1835-1838 (1987)
David D Díaz et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 12(9), 2593-2606 (2006-01-10)
A highly regio- and stereoselective intramolecular [1,5]-hydrogen-transfer process is described. Treatment of gamma-benzyl-protected Co2(CO)6-alpha,gamma-acetylenic diols with BF3 x OEt2 provides bis-homopropargylic alcohols. The reaction occurs within seconds, tolerates a wide range of functionalities, and provides good yields. When the ether
Shengli Zhu et al.
Journal of the mechanical behavior of biomedical materials, 13, 166-173 (2012-08-18)
We developed Ni- and Be-free Zr(45+x)Cu(40-x)Al₇Pd₅Nb₃ bulk metallic glasses with large glass-forming ability and unusual plasticity. The alloys have large critical diameters (larger than 10 mm) in a wide composition range (x=0-20). The Zr₅₀Cu₃₅Al₇Pd₅Nb₃ and Zr₅₅Cu₃₀Al₇Pd₅Nb₃ alloys exhibit the largest
Mark Niemeyer et al.
Inorganic chemistry, 36(21), 4688-4696 (2001-10-24)
The use of the terphenyl substituent -C(6)H(3)-2,6-Mes(2) (abbreviated Ar) has permitted the synthesis of several new low-coordinate beryllium compounds. Reaction of 1 equiv of LiAr with BeCl(2)(OEt(2))(2) or BeBr(2)(OEt(2))(2) (1) gives the monomeric complexes ArBeX(OEt(2)) (X = Cl (2); Br

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service