Skip to Content
Merck
All Photos(1)

Key Documents

377767

Sigma-Aldrich

Martius Yellow

Dye content 85 %

Synonym(s):

2,4-Dinitro-1-naphthol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(O2N)2C10H5OH
CAS Number:
Molecular Weight:
234.17
Beilstein:
2057462
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

form

powder

composition

Dye content, 85%

mp

130-133 °C (lit.)

solubility

methanol: 1 mg/mL

λmax

430 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Oc1c(cc([N+]([O-])=O)c2ccccc12)[N+]([O-])=O

InChI

1S/C10H6N2O5/c13-10-7-4-2-1-3-6(7)8(11(14)15)5-9(10)12(16)17/h1-5,13H

InChI key

FFRBMBIXVSCUFS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Sabnis RW
Handbook of Acid-Base Indicators, 215-215 (2007)
Entamoeba histolytica: demonstration by PAS/Martius yellow technique.
G Hulman et al.
Medical laboratory sciences, 44(4), 396-397 (1987-10-01)
Sethi A
Systematic Lab Experiments in Organic Chemistry, 770-770 (2003)
D Roberts et al.
Langmuir : the ACS journal of surfaces and colloids, 28(46), 16196-16206 (2012-10-24)
The effect of peptide charge on the self-assembly, gelation behavior, and model drug release profiles has been explored here for three octa-peptides, VEVKVEVK (VEK2), VKVKVEVK (VEK3), and VEVEVKVE (VEK1), that carry a net charge of 0, +2, and -2 at

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service