Skip to Content
Merck
All Photos(3)

Key Documents

377457

Sigma-Aldrich

Manganese(III) oxide

−325 mesh, 99%

Synonym(s):

Manganese sesquioxide

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$198.00
1 KG
$613.00

$198.00


Estimated to ship onMarch 26, 2025


Request a Bulk Order

Select a Size

Change View
250 G
$198.00
1 KG
$613.00

About This Item

Linear Formula:
Mn2O3
CAS Number:
Molecular Weight:
157.87
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

$198.00


Estimated to ship onMarch 26, 2025


Request a Bulk Order

Quality Level

Assay

99%

form

powder

particle size

−325 mesh

density

4.5 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

O=[Mn]O[Mn]=O

InChI

1S/2Mn.3O

InChI key

GEYXPJBPASPPLI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Precursor to LiMn2O4. A useful battery cathode.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Magnetic materials find diverse applications from data storage to renewable energy.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service