Skip to Content
Merck
All Photos(2)

Documents

376361

Sigma-Aldrich

(R)-α-Acryloyloxy-β,β-dimethyl-γ-butyrolactone

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12O4
CAS Number:
Molecular Weight:
184.19
Beilstein:
4668587
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

optical activity

[α]20/D +6.0°, c = 17 in methylene chloride

contains

0.1% monomethyl ether hydroquinone as stabilizer

refractive index

n20/D 1.46 (lit.)

bp

84 °C/0.1 mmHg (lit.)

mp

7-8 °C (lit.)

density

1.125 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

CC1(C)COC(=O)[C@@H]1OC(=O)C=C

InChI

1S/C9H12O4/c1-4-6(10)13-7-8(11)12-5-9(7,2)3/h4,7H,1,5H2,2-3H3/t7-/m0/s1

InChI key

ICMBTQSRUNSRKI-ZETCQYMHSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service