Skip to Content
Merck
All Photos(1)

Key Documents

375365

Sigma-Aldrich

4-Methyl-2-phenylimidazole

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10N2
CAS Number:
Molecular Weight:
158.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
95%

Assay

95%

mp

180-183 °C (lit.)

SMILES string

Cc1c[nH]c(n1)-c2ccccc2

InChI

1S/C10H10N2/c1-8-7-11-10(12-8)9-5-3-2-4-6-9/h2-7H,1H3,(H,11,12)

InChI key

TYOXIFXYEIILLY-UHFFFAOYSA-N

General description

4-Methyl-2-phenylimidazole is a 4-substituted-2-phenylimidazole.[1] It participates in ring-opening of various bicyclic olefins to afford trans-3,4-disubstituted derivatives.[2] The FT-IR, FT-Raman and FT-NMR spectra of 4-methyl-2-phenylimidazole has been recorded.[3]

Application

4-Methyl-2-phenylimidazole is suitable for use to investgate the cytotoxic activity of 4-trifluoromethylimidazoles against human tumor and normal cells.[2] It may be used in following studies:
  • Curing of epoxy system consisting of a diglycidyl ether of bisphenol-F epoxy resin, 1,4-butanediol.[1]
  • Curing of bisphenol-F epoxy resin.[4]
  • Synthesis of aza-heterocyclic compounds.[5]
  • Preparation of 1-[(2-chlorophenyl)(diphenyl)methyl]-4-methyl-2-phenyl-1H-imidazole.[6]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rhodium (III)-catalyzed CH activation of phenylazoles toward CN bond cleavage of diazabicyclic olefins: A facile access to mono-and biscyclopentenyl-functionalized aza-heteroaromatics
Praveen P, et al.
Synlett, 25, 0275-0279 (2014)
Fumihiro Takekawa et al.
Anticancer research, 27(6B), 4065-4069 (2008-01-30)
Fourteen 4-trifluoromethylimidazole derivatives were investigated for their cytotoxicity against three human normal cells (gingival fibroblast HGF, pulp cell HPC and periodontal ligament fibroblast HPLF) and four human tumor cell lines (oral squamous cell carcinoma HSC-2, HSC-3, HSC-4 and promyelocytic leukemia
Physical properties of a bisphenol-F epoxy containing a silica filler treated with silane coupling agents.
Case SL and Ward TC.
Journal of Adhesion, 79(2), 105-121 (2003)
Cure profiles, crosslink density, residual stresses, and adhesion in a model epoxy.
Case SL, et al.
Polymer, 46(24), 1083-1040 (2005)
Theoretical investigations on the molecular structure and vibrational spectral analysis of 4-methyl 2-phenylimidazole.
Erdogdu Y, et al.
Optics and Spectroscopy, 113(6), 596-606 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service