Skip to Content
Merck
All Photos(1)

Key Documents

375322

Sigma-Aldrich

Ethyl 2-oxo-4-phenylbutyrate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH2COCOOCH2CH3
CAS Number:
Molecular Weight:
206.24
Beilstein:
2725083
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.504 (lit.)

bp

132 °C/2 mmHg (lit.)

density

1.091 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)C(=O)CCc1ccccc1

InChI

1S/C12H14O3/c1-2-15-12(14)11(13)9-8-10-6-4-3-5-7-10/h3-7H,2,8-9H2,1H3

InChI key

STPXIOGYOLJXMZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl 2-oxo-4-phenylbutyrate is an aliphatic α-ketoester. Bioreduction of ethyl 2-oxo-4-phenylbutyrate is reported to yield ethyl (R)-2-hydroxy-4-phenylbutanoate. The effect of ionic liquid on the asymmetric reduction of ethyl 2-oxo-4-phenylbutyrate by Saccharomyces cerevisiae has been reported. Asymmetric reduction of ethyl 2-oxo-4-phenylbutyrate using a bacterial reductase is reported. Enantioselective hydrogenation of ethyl 2-oxo-4-phenylbutyrate using homogeneous Rh-diphosphine and heterogeneous Pt/Al2O3-cinchona catalysts has been reported.

Application

Ethyl 2-oxo-4-phenylbutyrate may be used in the synthesis of ethyl (R)-2-hydroxy-4-phenylbutyrate, an important chiral precursor for angiotensin-converting enzyme (ACE) inhibitor.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Efficient Reduction of Ethyl 2-Oxo-4-phenylbutyrate at 620 g? L- 1 by a Bacterial Reductase with Broad Substrate Spectrum.
Ni Y, et al.
J. Mol. Catal. A: Chem., 107(1), 85-94 (1996)
Yu-Gang Shi et al.
Journal of industrial microbiology & biotechnology, 35(11), 1419-1424 (2008-08-22)
The effect of ionic liquid 1-butyl-3-methylimidazolium hexafluorophosphate ([BMIM][PF6]) on the asymmetric reduction of ethyl 2-oxo-4-phenylbutyrate (EOPB) to synthesize optical active ethyl 2-hydroxy-4-phenylbutyrate (EHPB) catalyzed by Saccharomyces cerevisiae was investigated. (R)-EHPB [70.4%, e.e.(R)] is obtained using ethyl ether or benzene as
Ye Ni et al.
Journal of biotechnology, 168(4), 493-498 (2013-10-15)
Ethyl (R)-2-hydroxy-4-phenylbutanoate [(R)-HPBE] is a versatile and important chiral intermediate for the synthesis of angiotensin-converting enzyme (ACE) inhibitors. Recombinant E. coli strain coexpressing a novel NADPH-dependent carbonyl reductase gene iolS and glucose dehydrogenase gene gdh from Bacillus subtilis showed excellent
Nai-Dong Shen et al.
Organic letters, 14(8), 1982-1985 (2012-04-07)
A new reductase, CgKR2, with the ability to reduce ethyl 2-oxo-4-phenylbutyrate (OPBE) to ethyl (R)-2-hydroxy-4-phenylbutyrate ((R)-HPBE), an important chiral precursor for angiotensin-converting enzyme (ACE) inhibitors, was discovered. For the first time, (R)-HPBE with >99% ee was produced via bioreduction of
Efficient Reduction of Ethyl 2-Oxo-4-phenylbutyrate at 620 g? L- 1 by a Bacterial Reductase with Broad Substrate Spectrum.
Ni Y, et al.
Advanced Synthesis & Catalysis, 353(8), 1213-1217 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service