Skip to Content
Merck
All Photos(1)

Key Documents

375233

Sigma-Aldrich

Dimethyl 3-methylglutarate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(CH2CO2CH3)2
CAS Number:
Molecular Weight:
174.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.425 (lit.)

bp

110 °C/19 mmHg (lit.)

density

1.052 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

COC(=O)CC(C)CC(=O)OC

InChI

1S/C8H14O4/c1-6(4-7(9)11-2)5-8(10)12-3/h6H,4-5H2,1-3H3

InChI key

YIJLMTNDXYVGPQ-UHFFFAOYSA-N

General description

Dimethyl 3-methylglutarate is an ester. It participates in the synthesis of (R,Z)- muscenone, a valuable perfume ingredient.

Application

Building block for chemoenzymatic asymmetric synthesis.
Dimethyl 3-methylglutarate may be used in the synthesis of (R)- and (S)-4-amino-3-methylbutanoic acids, via initial enantioselective hydrolysis with pig liver esterase. It may be used in the preparation of optically active form of verrucarinic acid derivative. It may be used as building block for chemoenzymatic asymmetric synthesis.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

206.6 °F - closed cup

Flash Point(C)

97.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of Verrucarin A and 3a-Hydroxyverrucarin A from Verrucarol and Diacetoxyscripenol (Anguidine). 39th Communication on Verrucarins and Roridins.
Mohr P, et al.
Helvetica Chimica Acta, 65(5), 1412-1417 (1982)
An efficient route to the musk odorant (< i> R</i>,< i> Z</i>)-5-muscenone via base-metal-catalysis.
Lehr K and Furstner A.
Tetrahedron, 68(37), 7695-7700 (2012)
Chemoenzymatic synthesis of (R)-and (S)-4-amino-3-methylbutanoic acids.
Andruszkiewicz R, et al.
Synthetic Communications, 20, 159-166 (1990)
Tetrahedron, 44, 1477-1477 (1988)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service